3-(Benzyloxy)pyridine-4-carbonitrile 97%

Lieferant: Apollo Scientific
Achtung

OR33059-1G
APOSOR33059-1GEA 0 EUR
APOSOR33059-1G
3-(Benzyloxy)pyridine-4-carbonitrile 97%
3-(Benzyloxy)pyridine-4-carbonitrile
Formel: C₁₃H₁₀N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD23617692
CAS-Nummer: 78790-76-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR