4-Benzyl-1(2H)-phthalazinone 95%
Lieferant: Apollo Scientific
Synonyme:
4-Benzylphthalazin-1(2H)-one
OR33063-1G
OR33063-5G
OR33063-25G
APOSOR33063-1GEA
180
EUR
APOSOR33063-1G
APOSOR33063-5G
APOSOR33063-25G
4-Benzyl-1(2H)-phthalazinone 95%
4-Benzyl-1(2H)-phthalazinone
Formel:
C₁₅H₁₂N₂O Molecular Weight: 236.28 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00139376 CAS-Nummer: 32003-14-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...