3-(2-Phenylhydrazin-1-ylidene)pentane-2,4-dione 95%

Lieferant: Apollo Scientific

OR33064-1G
APOSOR33064-1GEA 427 EUR
APOSOR33064-1G
3-(2-Phenylhydrazin-1-ylidene)pentane-2,4-dione 95%
3-(2-Phenylhydrazin-1-ylidene)pentane-2,4-dione
Formel: C₁₁H₁₂N₂O₂
Molecular Weight: 204,225 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00459047
CAS-Nummer: 6134-57-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR