3-[(1E)-(Dimethylamino)methylidene]-1-phenylthiourea, tech

Lieferant: Apollo Scientific

OR33065-1G
APOSOR33065-1GEA 1570 EUR
APOSOR33065-1G
3-[(1E)-(Dimethylamino)methylidene]-1-phenylthiourea, tech
3-[(1E)-(Dimethylamino)methylidene]-1-phenylthiourea
Formel: C₁₀H₁₃N₃S
Molecular Weight: 207,295 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00139354
CAS-Nummer: 1661847-00-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR