6-[(4-Methylphenyl)methyl]pyridazin-3-ol, tech

Lieferant: Apollo Scientific

OR33068-1G
APOSOR33068-1GEA 1570 EUR
APOSOR33068-1G
6-[(4-Methylphenyl)methyl]pyridazin-3-ol, tech
6-[(4-Methylphenyl)methyl]pyridazin-3-ol
Formel: C₁₂H₁₂N₂O
Molecular Weight: 200,236 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00214782
CAS-Nummer: 339008-40-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR