(3Z)-3-{[(2-Hydroxyphenyl)amino]methylidene}-1,3-dihydro-2-benzofuran-1-one, tech

Lieferant: Apollo Scientific

OR33069-1G
APOSOR33069-1GEA 1570 EUR
APOSOR33069-1G
(3Z)-3-{[(2-Hydroxyphenyl)amino]methylidene}-1,3-dihydro-2-benzofuran-1-one, tech
(3Z)-3-{[(2-Hydroxyphenyl)amino]methylidene}-1,3-dihydro-2-benzofuran-1-one
Formel: C₁₅H₁₁NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00139664
CAS-Nummer: 339008-47-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR