1-[(4-Chlorophenyl)methyl]-6-oxo-1,6-dihydropyridine-3-carboxylic acid, tech
Lieferant: Apollo Scientific
OR33071-1G
APOSOR33071-1GEA
1570
EUR
APOSOR33071-1G
1-[(4-Chlorophenyl)methyl]-6-oxo-1,6-dihydropyridine-3-carboxylic acid, tech
1-[(4-Chlorophenyl)methyl]-6-oxo-1,6-dihydropyridine-3-carboxylic acid
Formel:
C₁₃H₁₀ClNO₃ Molecular Weight: 263,676 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00139926 CAS-Nummer: 339008-74-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...