5-Amino-1-[(3,4-dichlorophenyl)methyl]-1,2-dihydropyridin-2-one 95%

Lieferant: Apollo Scientific

OR33074-1G
APOSOR33074-1GEA 1330 EUR
APOSOR33074-1G
5-Amino-1-[(3,4-dichlorophenyl)methyl]-1,2-dihydropyridin-2-one 95%
5-Amino-1-[(3,4-dichlorophenyl)methyl]-1,2-dihydropyridin-2-one
Formel: C₁₂H₁₀Cl₂N₂O
Molecular Weight: 269,127 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00139933
CAS-Nummer: 339009-11-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR