(2Z)-2-(2,4-Dichlorophenyl)-3-hydroxyprop-2-enenitrile, tech

Lieferant: Apollo Scientific

OR33075-1G
APOSOR33075-1GEA 1570 EUR
APOSOR33075-1G
(2Z)-2-(2,4-Dichlorophenyl)-3-hydroxyprop-2-enenitrile, tech
(2Z)-2-(2,4-Dichlorophenyl)-3-hydroxyprop-2-enenitrile
Formel: C₉H₅Cl₂NO
Molecular Weight: 214,048 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00141627
CAS-Nummer: 115716-98-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR