2-Methylpyrazolo[1,5-a]pyrimidine-6-carbonitrile 95%

Lieferant: Apollo Scientific

OR33076-1G
APOSOR33076-1GEA 671 EUR
APOSOR33076-1G
2-Methylpyrazolo[1,5-a]pyrimidine-6-carbonitrile 95%
2-Methylpyrazolo[1,5-a]pyrimidine-6-carbonitrile
Formel: C₈H₆N₄
Molecular Weight: 158,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00173619
CAS-Nummer: 339010-67-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR