Methyl 6-(4-phenylpiperazin-1-yl)pyridine-3-carboxylate, tech

Lieferant: Apollo Scientific

OR33078-1G
APOSOR33078-1GEA 1570 EUR
APOSOR33078-1G
Methyl 6-(4-phenylpiperazin-1-yl)pyridine-3-carboxylate, tech
Methyl 6-(4-phenylpiperazin-1-yl)pyridine-3-carboxylate
Formel: C₁₇H₁₉N₃O₂
Molecular Weight: 297,352 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01316590
CAS-Nummer: 132521-73-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR