Methyl 4-chloro-1-(3-chlorophenyl)-6-oxo-1,6-dihydropyridazine-3-carboxylate, tech

Lieferant: Apollo Scientific

OR33079-1G
APOSOR33079-1GEA 1570 EUR
APOSOR33079-1G
Methyl 4-chloro-1-(3-chlorophenyl)-6-oxo-1,6-dihydropyridazine-3-carboxylate, tech
Methyl 4-chloro-1-(3-chlorophenyl)-6-oxo-1,6-dihydropyridazine-3-carboxylate
Formel: C₁₂H₈Cl₂N₂O₃
Molecular Weight: 299,11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03305735
CAS-Nummer: 866051-44-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR