8-Methyl-N-phenyl-1-oxa-4,8-diazaspiro[4.5]decane-4-carboxamide, tech

Lieferant: Apollo Scientific

OR33080-1G
APOSOR33080-1GEA 1570 EUR
APOSOR33080-1G
8-Methyl-N-phenyl-1-oxa-4,8-diazaspiro[4.5]decane-4-carboxamide, tech
8-Methyl-N-phenyl-1-oxa-4,8-diazaspiro[4.5]decane-4-carboxamide
Formel: C₁₅H₂₁N₃O₂
Molecular Weight: 275,346 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00172862
CAS-Nummer: 339011-64-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR