(2Z)-3-(4-Chlorophenyl)-2-[(1E)-(methoxyimino)methyl]prop-2-enenitrile, tech

Lieferant: Apollo Scientific

OR33081-1G
APOSOR33081-1GEA 1570 EUR
APOSOR33081-1G
(2Z)-3-(4-Chlorophenyl)-2-[(1E)-(methoxyimino)methyl]prop-2-enenitrile, tech
(2Z)-3-(4-Chlorophenyl)-2-[(1E)-(methoxyimino)methyl]prop-2-enenitrile
Formel: C₁₁H₉ClN₂O
Molecular Weight: 220,655 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00663738
CAS-Nummer: 339011-91-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR