4-Acetyl-1-methyl-1H-pyrrole-2-carboxylic acid 95%
Lieferant: Apollo Scientific
OR33082-1G
APOSOR33082-1GEA
488
EUR
APOSOR33082-1G
4-Acetyl-1-methyl-1H-pyrrole-2-carboxylic acid 95%
4-Acetyl-1-methyl-1H-pyrrole-2-carboxylic acid
Formel:
C₈H₉NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00665069 CAS-Nummer: 339011-93-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...