3-[4-(2-Chlorophenyl)piperazin-1-yl]-3-iminopropanenitrile, tech

Lieferant: Apollo Scientific

OR33085-1G
APOSOR33085-1GEA 1570 EUR
APOSOR33085-1G
3-[4-(2-Chlorophenyl)piperazin-1-yl]-3-iminopropanenitrile, tech
3-[4-(2-Chlorophenyl)piperazin-1-yl]-3-iminopropanenitrile
Formel: C₁₃H₁₅ClN₄
Molecular Weight: 262,738 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00664799
CAS-Nummer: 344276-15-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR