2-{[(3-Methoxyphenyl)amino]methylidene}cyclohexane-1,3-dione, tech
Lieferant: Apollo Scientific
OR33087-1G
APOSOR33087-1GEA
1570
EUR
APOSOR33087-1G
2-{[(3-Methoxyphenyl)amino]methylidene}cyclohexane-1,3-dione, tech
2-{[(3-Methoxyphenyl)amino]methylidene}cyclohexane-1,3-dione
Formel:
C₁₄H₁₅NO₃ Molecular Weight: 245,274 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00793089 CAS-Nummer: 55388-67-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...