(3Z)-3-{[Hydroxy(methyl)amino]methylidene}-2,3-dihydro-1H-indol-2-one, tech

Lieferant: Apollo Scientific

OR33088-1G
APOSOR33088-1GEA 1570 EUR
APOSOR33088-1G
(3Z)-3-{[Hydroxy(methyl)amino]methylidene}-2,3-dihydro-1H-indol-2-one, tech
(3Z)-3-{[Hydroxy(methyl)amino]methylidene}-2,3-dihydro-1H-indol-2-one
Formel: C₁₀H₁₀N₂O₂
Molecular Weight: 190,199 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00232406
CAS-Nummer: 1164473-20-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR