1-[2,3-Dichloro-4-(ethylsulphonyl)phenyl]-1-ethanone 95%

Lieferant: Apollo Scientific

OR33091-1G
APOSOR33091-1GEA 671 EUR
APOSOR33091-1G
1-[2,3-Dichloro-4-(ethylsulphonyl)phenyl]-1-ethanone 95%
1-[2,3-Dichloro-4-(ethylsulphonyl)phenyl]-1-ethanone
Formel: C₁₀H₁₀Cl₂O₃S
Molecular Weight: 281,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01935964
CAS-Nummer: 338982-46-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR