2-(Methylsulphanyl)-5-phenylpyrimidine 95%
Lieferant: Apollo Scientific
OR33092-1G
APOSOR33092-1GEA
387
EUR
APOSOR33092-1G
2-(Methylsulphanyl)-5-phenylpyrimidine 95%
2-(Methylsulphanyl)-5-phenylpyrimidine
Formel:
C₁₁H₁₀N₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD00204407 CAS-Nummer: 31408-20-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...