6-Methyl-3-phenyl-1,2,4-triazin-5-ol, tech

Lieferant: Apollo Scientific

OR33094-1G
APOSOR33094-1GEA 1570 EUR
APOSOR33094-1G
6-Methyl-3-phenyl-1,2,4-triazin-5-ol, tech
6-Methyl-3-phenyl-1,2,4-triazin-5-ol
Formel: C₁₀H₉N₃O
Molecular Weight: 187,198 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01315359
CAS-Nummer: 33449-34-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR