3-[2-(1H-Pyrrol-1-yl)phenoxy]butan-2-one, tech
Lieferant: Apollo Scientific
OR33100-1G
APOSOR33100-1GEA
1570
EUR
APOSOR33100-1G
3-[2-(1H-Pyrrol-1-yl)phenoxy]butan-2-one, tech
3-[2-(1H-Pyrrol-1-yl)phenoxy]butan-2-one
Formel:
C₁₄H₁₅NO₂ Molecular Weight: 229,274 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00792993 CAS-Nummer: 339014-73-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...