Dimethyl[(E)-2-(1-nitronaphthalen-2-yl)ethenyl]amine, tech

Lieferant: Apollo Scientific

OR33102-1G
APOSOR33102-1GEA 1570 EUR
APOSOR33102-1G
Dimethyl[(E)-2-(1-nitronaphthalen-2-yl)ethenyl]amine, tech
Dimethyl[(E)-2-(1-nitronaphthalen-2-yl)ethenyl]amine
Formel: C₁₄H₁₄N₂O₂
Molecular Weight: 242,273 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00664614
CAS-Nummer: 176853-40-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR