(2E)-2-[(Furan-2-yl)methylidene]-1-azabicyclo[2.2.2]octan-3-one, tech

Lieferant: Apollo Scientific

OR33105-1G
APOSOR33105-1GEA 1570 EUR
APOSOR33105-1G
(2E)-2-[(Furan-2-yl)methylidene]-1-azabicyclo[2.2.2]octan-3-one, tech
(2E)-2-[(Furan-2-yl)methylidene]-1-azabicyclo[2.2.2]octan-3-one
Formel: C₁₂H₁₃NO₂
Molecular Weight: 203,237 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00219029
CAS-Nummer: 82955-18-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR