1-(4-Acetylphenyl)-3,3-dimethylazetidin-2-one, tech
Lieferant: Apollo Scientific
OR33107-1G
APOSOR33107-1GEA
1570
EUR
APOSOR33107-1G
1-(4-Acetylphenyl)-3,3-dimethylazetidin-2-one, tech
1-(4-Acetylphenyl)-3,3-dimethylazetidin-2-one
Formel:
C₁₃H₁₅NO₂ Molecular Weight: 217,264 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00793414 CAS-Nummer: 339015-93-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...