3-Phenyl-1-(phenylamino)-2,5-dihydro-1H-pyrrole-2,5-dione, tech

Lieferant: Apollo Scientific

OR33111-1G
APOSOR33111-1GEA 1570 EUR
APOSOR33111-1G
3-Phenyl-1-(phenylamino)-2,5-dihydro-1H-pyrrole-2,5-dione, tech
3-Phenyl-1-(phenylamino)-2,5-dihydro-1H-pyrrole-2,5-dione
Formel: C₁₆H₁₂N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00974736
CAS-Nummer: 49811-67-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR