3-(4-Methoxyphenyl)furan-2,5-dione 95%

Lieferant: Apollo Scientific

OR33113-1G
APOSOR33113-1GEA 874 EUR
APOSOR33113-1G
3-(4-Methoxyphenyl)furan-2,5-dione 95%
3-(4-Methoxyphenyl)furan-2,5-dione
Formel: C₁₁H₈O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00974742
CAS-Nummer: 4665-00-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR