3-(4-Methoxyphenyl)-1-(phenylamino)-2,5-dihydro-1H-pyrrole-2,5-dione, tech

Lieferant: Apollo Scientific

OR33114-1G
APOSOR33114-1GEA 1570 EUR
APOSOR33114-1G
3-(4-Methoxyphenyl)-1-(phenylamino)-2,5-dihydro-1H-pyrrole-2,5-dione, tech
3-(4-Methoxyphenyl)-1-(phenylamino)-2,5-dihydro-1H-pyrrole-2,5-dione
Formel: C₁₇H₁₄N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00974747
CAS-Nummer: 49811-72-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR