3-(4-Chlorophenyl)-6-methyl-1,2,4-triazin-5-ol, tech

Lieferant: Apollo Scientific

OR33115-1G
APOSOR33115-1GEA 1570 EUR
APOSOR33115-1G
3-(4-Chlorophenyl)-6-methyl-1,2,4-triazin-5-ol, tech
3-(4-Chlorophenyl)-6-methyl-1,2,4-triazin-5-ol
Formel: C₁₀H₈ClN₃O
Molecular Weight: 221,643 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05668805
CAS-Nummer: 126542-93-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR