3-(3-Chlorophenyl)-6-methyl-1,2,4-triazin-5-ol 95%

Lieferant: Apollo Scientific

OR33116-1G
APOSOR33116-1GEA 671 EUR
APOSOR33116-1G
3-(3-Chlorophenyl)-6-methyl-1,2,4-triazin-5-ol 95%
3-(3-Chlorophenyl)-6-methyl-1,2,4-triazin-5-ol
Formel: C₁₀H₈ClN₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD04117818
CAS-Nummer: 886360-72-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR