(2E)-1-(2,5-Dichlorothiophen-3-yl)-3-(dimethylamino)prop-2-en-1-one, tech
Lieferant: Apollo Scientific
OR33124-1G
APOSOR33124-1GEA
1570
EUR
APOSOR33124-1G
(2E)-1-(2,5-Dichlorothiophen-3-yl)-3-(dimethylamino)prop-2-en-1-one, tech
(2E)-1-(2,5-Dichlorothiophen-3-yl)-3-(dimethylamino)prop-2-en-1-one
Formel:
C₉H₉Cl₂NOS Lagertemperatur: Raumtemperatur |
MDL:
MFCD01443682 CAS-Nummer: 166196-79-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...