(E)-N-Benzyl-2-phenylethene-1-sulphonamide, tech

Lieferant: Apollo Scientific

OR33129-1G
APOSOR33129-1GEA 1570 EUR
APOSOR33129-1G
(E)-N-Benzyl-2-phenylethene-1-sulphonamide, tech
(E)-N-Benzyl-2-phenylethene-1-sulphonamide
Formel: C₁₅H₁₅NO₂S
Molecular Weight: 273,35 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00222948
CAS-Nummer: 13719-49-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR