(2E)-3-(Dimethylamino)-1-[4-(pyrimidin-2-yloxy)phenyl]prop-2-en-1-one, tech

Lieferant: Apollo Scientific

OR33131-1G
APOSOR33131-1GEA 1570 EUR
APOSOR33131-1G
(2E)-3-(Dimethylamino)-1-[4-(pyrimidin-2-yloxy)phenyl]prop-2-en-1-one, tech
(2E)-3-(Dimethylamino)-1-[4-(pyrimidin-2-yloxy)phenyl]prop-2-en-1-one
Formel: C₁₅H₁₅N₃O₂
Molecular Weight: 269.299 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01443774
CAS-Nummer: 339018-66-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR