Ethyl 2-(4-methyl-2-oxo-1,2-dihydroquinolin-1-yl)acetate, tech

Lieferant: Apollo Scientific

OR33132-1G
APOSOR33132-1GEA 1570 EUR
APOSOR33132-1G
Ethyl 2-(4-methyl-2-oxo-1,2-dihydroquinolin-1-yl)acetate, tech
Ethyl 2-(4-methyl-2-oxo-1,2-dihydroquinolin-1-yl)acetate
Formel: C₁₄H₁₅NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD01444199
CAS-Nummer: 103368-18-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR