Ethyl 2-(4,8-dimethyl-2-oxo-1,2-dihydroquinolin-1-yl)acetate, tech
Lieferant: Apollo Scientific
OR33133-1G
APOSOR33133-1GEA
1570
EUR
APOSOR33133-1G
Ethyl 2-(4,8-dimethyl-2-oxo-1,2-dihydroquinolin-1-yl)acetate, tech
Ethyl 2-(4,8-dimethyl-2-oxo-1,2-dihydroquinolin-1-yl)acetate
Formel:
C₁₅H₁₇NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD01444200 CAS-Nummer: 145667-25-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...