(1-Methyl-1H-imidazol-5-yl)methyl 4-chlorobenzoate, tech

Lieferant: Apollo Scientific

OR33134-1G
APOSOR33134-1GEA 1570 EUR
APOSOR33134-1G
(1-Methyl-1H-imidazol-5-yl)methyl 4-chlorobenzoate, tech
(1-Methyl-1H-imidazol-5-yl)methyl 4-chlorobenzoate
Formel: C₁₂H₁₁ClN₂O₂
Molecular Weight: 250,681 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04125963
CAS-Nummer: 866131-15-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR