(2E)-3-Amino-3-(4-chlorophenyl)prop-2-enenitrile 95%

Lieferant: Apollo Scientific

OR33136-1G
APOSOR33136-1GEA 874 EUR
APOSOR33136-1G
(2E)-3-Amino-3-(4-chlorophenyl)prop-2-enenitrile 95%
(2E)-3-Amino-3-(4-chlorophenyl)prop-2-enenitrile
Formel: C₉H₇ClN₂
Molecular Weight: 178,618 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01871636
CAS-Nummer: 107840-43-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR