3-Amino-3-(4-bromophenyl)acrylonitrile 95%

Lieferant: Apollo Scientific

OR33137-1G OR33137-25G OR33137-5G
APOSOR33137-1GEA 180 EUR
APOSOR33137-1G APOSOR33137-25G APOSOR33137-5G
3-Amino-3-(4-bromophenyl)acrylonitrile 95%
3-Amino-3-(4-bromophenyl)acrylonitrile
Formel: C₉H₇BrN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01871637
CAS-Nummer: 748807-82-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR