N-(2,3-Dimethylphenyl)acetamide 95%

Lieferant: Apollo Scientific

Synonyme: 2,3-Dimethylacetanilide

OR33138-1G
APOSOR33138-1GEA 427 EUR
APOSOR33138-1G
N-(2,3-Dimethylphenyl)acetamide 95%
N-(2,3-Dimethylphenyl)acetamide
Formel: C₁₀H₁₃NO
Molecular Weight: 163,216 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00130064
CAS-Nummer: 134-98-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR