2-Chlor-5-hydroxyphenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (2-Chloro-5-hydroxyphenyl)boronic acid

OR3314-100MG
APOSOR3314-100MGEA 21.9 EUR
APOSOR3314-100MG
2-Chlor-5-hydroxyphenylboronsäure 98%
2-Chlor-5-hydroxyphenylboronsäure
Formel: C₆H₆BClO₃
Molecular Weight: 172.3700 g/mol
Schmelzpunkt: 263…265 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08689478
CAS-Nummer: 913835-71-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR