N-(2,5-Dimethylphenyl)acetamide 95%

Lieferant: Apollo Scientific

OR33140-1G
APOSOR33140-1GEA 440 EUR
APOSOR33140-1G
N-(2,5-Dimethylphenyl)acetamide 95%
N-(2,5-Dimethylphenyl)acetamide
Formel: C₁₀H₁₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD00220610
CAS-Nummer: 2050-44-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR