(3Z)-3-[(Phenylamino)methylidene]-3,4-dihydro-2H-1-benzopyran-2,4-dione, tech

Lieferant: Apollo Scientific

OR33141-1G
APOSOR33141-1GEA 1570 EUR
APOSOR33141-1G
(3Z)-3-[(Phenylamino)methylidene]-3,4-dihydro-2H-1-benzopyran-2,4-dione, tech
(3Z)-3-[(Phenylamino)methylidene]-3,4-dihydro-2H-1-benzopyran-2,4-dione
Formel: C₁₆H₁₁NO₃
Molecular Weight: 265,263 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00100852
CAS-Nummer: 71592-11-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR