Methyl 2-(2,5-dimethyl-1H-pyrrol-1-yl)thiophene-3-carboxylate, tech

Lieferant: Apollo Scientific

OR33142-1G
APOSOR33142-1GEA 1570 EUR
APOSOR33142-1G
Methyl 2-(2,5-dimethyl-1H-pyrrol-1-yl)thiophene-3-carboxylate, tech
Methyl 2-(2,5-dimethyl-1H-pyrrol-1-yl)thiophene-3-carboxylate
Formel: C₁₂H₁₃NO₂S
Molecular Weight: 235,302 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01312398
CAS-Nummer: 478077-85-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR