6-Chloro-2,5-dimethylpyrazolo[1,5-a]pyrimidin-7-ol, tech

Lieferant: Apollo Scientific

OR33143-1G
APOSOR33143-1GEA 1570 EUR
APOSOR33143-1G
6-Chloro-2,5-dimethylpyrazolo[1,5-a]pyrimidin-7-ol, tech
6-Chloro-2,5-dimethylpyrazolo[1,5-a]pyrimidin-7-ol
Formel: C₈H₈ClN₃O
Molecular Weight: 197,622 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02570845
CAS-Nummer: 478077-86-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR