3-(4-Methoxyphenyl)-1-(pyridin-2-yl)propan-1-ol, tech
Lieferant: Apollo Scientific
OR33144-1G
APOSOR33144-1GEA
1570
EUR
APOSOR33144-1G
3-(4-Methoxyphenyl)-1-(pyridin-2-yl)propan-1-ol, tech
3-(4-Methoxyphenyl)-1-(pyridin-2-yl)propan-1-ol
Formel:
C₁₅H₁₇NO₂ Molecular Weight: 243,301 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02570997 CAS-Nummer: 478078-18-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...