N-(2-Chloroethyl)-1H,2H-naphtho[2,1-b]furan-2-carboxamide, tech
Lieferant: Apollo Scientific
OR33145-1G
APOSOR33145-1GEA
1570
EUR
APOSOR33145-1G
N-(2-Chloroethyl)-1H,2H-naphtho[2,1-b]furan-2-carboxamide, tech
N-(2-Chloroethyl)-1H,2H-naphtho[2,1-b]furan-2-carboxamide
Formel:
C₁₅H₁₄ClNO₂ Molecular Weight: 275,73 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02570920 CAS-Nummer: 478078-36-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...