tert-Butyl-2-(4-hydroxyphenoxy)acetate 95%
Lieferant: Apollo Scientific
OR33146-1G
APOSOR33146-1GEA
1070
EUR
APOSOR33146-1G
tert-Butyl-2-(4-hydroxyphenoxy)acetate 95%
tert-Butyl-2-(4-hydroxyphenoxy)acetate
Formel:
C₁₂H₁₆O₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD12025831 CAS-Nummer: 42806-92-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...