Methyl-3-amino-3-(4-bromophenyl)propanoate 95%

Lieferant: Apollo Scientific

OR33147-1G
APOSOR33147-1GEA 538 EUR
APOSOR33147-1G
Methyl-3-amino-3-(4-bromophenyl)propanoate 95%
Methyl-3-amino-3-(4-bromophenyl)propanoate
Formel: C₁₀H₁₂BrNO₂
Molecular Weight: 258,12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02286059
CAS-Nummer: 453557-71-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR