1-(4-Acetylphenyl)pyrrolidine-2,5-dione, tech

Lieferant: Apollo Scientific

OR33153-1G
APOSOR33153-1GEA 874 EUR
APOSOR33153-1G
1-(4-Acetylphenyl)pyrrolidine-2,5-dione, tech
1-(4-Acetylphenyl)pyrrolidine-2,5-dione
Formel: C₁₂H₁₁NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00452866
CAS-Nummer: 36254-39-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR