2-(3-Acetylphenyl)-2,3-dihydro-1H-isoindole-1,3-dione, tech

Lieferant: Apollo Scientific

OR33154-1G
APOSOR33154-1GEA 1570 EUR
APOSOR33154-1G
2-(3-Acetylphenyl)-2,3-dihydro-1H-isoindole-1,3-dione, tech
2-(3-Acetylphenyl)-2,3-dihydro-1H-isoindole-1,3-dione
Formel: C₁₆H₁₁NO₃
Molecular Weight: 265,263 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00628198
CAS-Nummer: 72801-61-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR